Assay Details
| Gene Symbol: LOC101928820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134633.1 | 2 - 3 | 250 | 80 | ||
| GenBank mRNA | AB371547.1 | - | 2 - 3 | 250 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | T-box 18 | TBX18 | CAKUT2 | -638.9kb | Sense | Chr. 6 - 84666834 - 84764236 on NCBI Build GRCh38 |
| Downstream | 5'-nucleotidase ecto | NT5E |
CALJA, CD73, E5NT, NT, NT5, NTE, eN, eNT |
45.2kb | Anti-sense | Chr. 6 - 85449584 - 85495791 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101928820
Genomic Map