Assay Details
| Gene Symbol: LOC105373100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134637.1 | 1 - 2 | 781 | 59 | ||
| GenBank mRNA | BC050343.1 | - | 1 - 2 | 751 | 59 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SH3 and multiple ankyrin repeat domains 3 | SHANK3 |
DEL22q13.3, PROSAP2, PSAP2, SCZD15, SPANK-2 |
-2.6kb | Anti-sense | Chr. 22 - 50674642 - 50733212 on NCBI Build GRCh38 |
| Downstream | acrosin | ACR | 55.0 | Anti-sense | Chr. 22 - 50738224 - 50745299 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105373100
Genomic Map