Assay Details
| Gene Symbol: LOC105372419 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134887.1 | 1 - 2 | 325 | 118 | ||
| GenBank mRNA | AL047654.1 | - | 1 - 2 | 118 | ||
| CN478580.1 | - | 1 - 2 | 118 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 296 | ZNF296 |
ZFP296, ZNF342 |
-8.9kb | Sense | Chr. 19 - 45071500 - 45076430 on NCBI Build GRCh38 |
| Downstream | protein phosphatase 1 regulatory subunit 37 | PPP1R37 | LRRC68 | 2.8kb | Anti-sense | Chr. 19 - 45093173 - 45147286 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105372419
Genomic Map