Assay Details
Gene Symbol: LOC105370014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_134998.1 | 2 - 3 | 182 | 90 | ||
GenBank mRNA | DB067934.1 | - | 2 - 3 | 182 | 90 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
SDS3 homolog, SIN3A corepressor complex component |
SUDS3 |
SAP45, SDS3 |
-340.7kb | Anti-sense | Chr. 12 - 118373189 - 118418035 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC105370016 | LOC105370016 | 58.7kb | Sense | Chr. 12 - 118818804 - 118833536 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
