Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010903.hg.1 |
ENST00000580703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012034.hg.1 |
ENST00000580337.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000548329.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000548329.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000548329.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000548329.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008921.hg.1 |
ENST00000578012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007437.hg.1 |
ENST00000581535.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012846.hg.1 |
ENST00000583422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007795.hg.1 |
ENST00000578350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007437.hg.1 |
ENST00000581535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008921.hg.1 |
ENST00000578012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000548329.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000548329.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000548329.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000548329.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012846.hg.1 |
ENST00000583422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010903.hg.1 |
ENST00000580703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007795.hg.1 |
ENST00000578350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012034.hg.1 |
ENST00000580337.1 |
Ensembl |