Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000548329.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000548329.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000548329.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011922.hg.1 |
ENST00000548329.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000546852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000548329.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000548329.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000548329.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011922.hg.1 |
ENST00000548329.1 |
Ensembl |