Assay Details
Gene Symbol: LINC01593 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_135071.1 | 1 - 2 | 127 | 78 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | solute carrier family 5 member 7 | SLC5A7 |
CHT, CHT1, HMN7A |
-35.1kb | Anti-sense | Chr. 2 - 107986514 - 108014497 on NCBI Build GRCh38 |
Downstream | long intergenic non-protein coding RNA 1594 | LINC01594 | TCONS_l2_00015893 | 115.0kb | Sense | Chr. 2 - 108167748 - 108217841 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
