Assay Details
Gene Symbol: LOC102724156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_135137.1 | 2 - 3 | 277 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | syncytin-2-like | LOC105369216 | -26.6kb | Sense | Chr. 9 - 88036710 - 88039819 on NCBI Build GRCh38 | |
Downstream | SPATA31 subfamily C member 2 | SPATA31C2 | FAM75C2 | 51.8kb | Sense | Chr. 9 - 88129305 - 88134985 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
