Assay Details
| Gene Symbol: LOC102724156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135138.1 | 1 - 2 | 177 | 64 | ||
| GenBank mRNA | BX377179.2 | - | 1 - 2 | 93 | 64 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | syncytin-2-like | LOC105369216 | -26.6kb | Sense | Chr. 9 - 88036710 - 88039819 on NCBI Build GRCh38 | |
| Downstream | SPATA31 subfamily C member 2 | SPATA31C2 | FAM75C2 | 51.4kb | Sense | Chr. 9 - 88129305 - 88134985 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC102724156
Genomic Map