Assay Details
Gene Symbol: LOC105376271 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_135145.1 | 1 - 2 | 170 | 95 | ||
GenBank mRNA | AA961598.1 | - | 1 - 2 | 94 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | protein phosphatase 6 catalytic subunit | PPP6C |
PP6, PP6C |
-9.6kb | Sense | Chr. 9 - 125146573 - 125189939 on NCBI Build GRCh38 |
Downstream | Rab9 effector protein with kelch motifs | RABEPK |
RAB9P40, bA65N13.1, p40 |
97.0 | Anti-sense | Chr. 9 - 125200493 - 125234159 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
