Assay Details
| Gene Symbol: LOC101928557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135193.1 | 1 - 2 | 184 | 104 | ||
| GenBank mRNA | AI393452.1 | - | 1 - 2 | 104 | ||
| BX118346.1 | - | 1 - 2 | 184 | 104 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 16 open reading frame 74 | C16orf74 | -41.3kb | Anti-sense | Chr. 16 - 85707517 - 85751083 on NCBI Build GRCh38 | |
| Downstream | cytochrome c oxidase subunit 4I1 | COX4I1 |
COX IV-1, COX4, COX4-1, COXIV, COXIV-1 |
6.6kb | Sense | Chr. 16 - 85799567 - 85807003 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101928557
Genomic Map