Assay Details
| Gene Symbol: GPR50-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135300.1 | 1 - 2 | 461 | 117 | ||
| GenBank mRNA | BX343514.2 | - | 1 - 2 | 462 | 117 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4330 | MIR4330 | -8.5kb | Anti-sense | Chr. X - 151168222 - 151168326 on NCBI Build GRCh38 | |
| Downstream | VMA21 vacuolar H+-ATPase homolog (S. cerevisiae) | VMA21 |
MEAX, XMEA |
218.7kb | Anti-sense | Chr. X - 151396555 - 151409364 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

GPR50-AS1
Genomic Map