Assay Details
Gene Symbol: LOC105370203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_135318.1 | 1 - 2 | 213 | 100 | ||
GenBank mRNA | CK903541.1 | - | 1 - 2 | 213 | 100 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | motilin receptor | MLNR |
GPR38, MTLR1 |
-13.1kb | Anti-sense | Chr. 13 - 49220338 - 49222377 on NCBI Build GRCh38 |
Downstream | cytidine and dCMP deaminase domain containing 1 | CDADC1 |
NYD-SP15, bA103J18.1 |
35.0 | Anti-sense | Chr. 13 - 49247875 - 49293486 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000594315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000594315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000594315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000594315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000594315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000594315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000594315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000594315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000595063.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000595063.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000595063.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000595063.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000595837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000595837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000595837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000595837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000595837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000595837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000595837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000595837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000597702.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000597702.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000597702.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000597702.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000597702.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000597702.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000598215.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000598215.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000598215.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000598215.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000598215.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000599801.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000599801.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000599801.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000599801.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000599801.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000599801.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000599801.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000599801.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000599801.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | ENST00000599801.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000588495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000594315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000594315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000594315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000594315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000594315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000594315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000594315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000594315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000595063.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000595063.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000595063.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000595063.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000595837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000595837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000595837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000595837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000595837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000595837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000595837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000595837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000597702.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000597702.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000597702.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000597702.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000597702.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000597702.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000598215.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000598215.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000598215.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000598215.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000598215.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000599801.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000599801.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000599801.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000599801.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000599801.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000599801.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000599801.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000599801.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000599801.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | ENST00000599801.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | NR_040109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010750.hg.1 | NR_040109.2 | Refseq |
