Assay Details
Gene Symbol: LOC102723345 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_135320.1 | 1 - 2 | 33 | 85 | ||
GenBank mRNA | BE464555.1 | - | 1 - 2 | 85 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | solute carrier family 7 member 1 | SLC7A1 |
ATRC1, CAT-1, ERR, HCAT1, REC1L |
-52.0kb | Anti-sense | Chr. 13 - 29509410 - 29595715 on NCBI Build GRCh38 |
Downstream | ubiquitin like 3 | UBL3 |
HCG-1, PNSC1 |
108.9kb | Anti-sense | Chr. 13 - 29764408 - 29850683 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
