Assay Details
Gene Symbol: LOC105377102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_135535.1 | 1 - 2 | 181 | 67 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ankyrin repeat and SOCS box containing 14 | ASB14 | Anti-sense | Chr. 3 - 57268351 - 57293047 on NCBI Build GRCh38 | ||
Downstream | dynein axonemal heavy chain 12 | DNAH12 |
DHC3, DLP12, DNAH12L, DNAH7L, DNAHC12, DNAHC3, DNHD2, HDHC3, HL-19, HL19 |
17.2kb | Anti-sense | Chr. 3 - 57293699 - 57545203 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
