Assay Details
| Gene Symbol: LOC101928403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135540.1 | 1 - 2 | 900 | 90 | ||
| GenBank mRNA | BC046476.1 | - | 1 - 2 | 900 | 90 | |
| CA432512.1 | - | 1 - 2 | 90 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | eukaryotic translation initiation factor 2 alpha kinase 3 | EIF2AK3 |
PEK, PERK, WRS |
-253.0 | Anti-sense | Chr. 2 - 88556740 - 88627576 on NCBI Build GRCh38 |
| Downstream | ribose 5-phosphate isomerase A | RPIA |
RPI, RPIAD |
59.8kb | Sense | Chr. 2 - 88691658 - 88750935 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101928403
Genomic Map