Assay Details
Gene Symbol: LOC102724714 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_135566.1 | 1 - 2 | 132 | 85 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | methyltransferase like 21A | METTL21A |
FAM119A, HCA557b, HSPA-KMT |
-36.5kb | Anti-sense | Chr. 2 - 207580631 - 207625928 on NCBI Build GRCh38 |
Downstream | cyclin Y like 1 | CCNYL1 | 44.0kb | Sense | Chr. 2 - 207711540 - 207756174 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
