Assay Details
| Gene Symbol: LOC101927635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135596.1 | 1 - 2 | 119 | 91 | ||
| GenBank mRNA | BG119374.1 | - | 1 - 2 | 119 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | FtsJ RNA methyltransferase homolog 1 (E. coli) | FTSJ1 |
CDLIV, JM23, MRX44, MRX9, SPB1, TRMT7 |
-20.2kb | Anti-sense | Chr. X - 48476021 - 48486364 on NCBI Build GRCh38 |
| Downstream | porcupine homolog (Drosophila) | PORCN |
DHOF, FODH, MG61, PORC, PPN |
115.0 | Anti-sense | Chr. X - 48508954 - 48520814 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101927635
Genomic Map