Assay Details
Gene Symbol: LOC105371766 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_135645.1 | 1 - 2 | 65 | 69 | ||
GenBank mRNA | BX103390.1 | - | 1 - 2 | 65 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
