Assay Details
Gene Symbol: LOC102723471 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_135669.1 | 3 - 4 | 395 | 86 | ||
GenBank mRNA | CB047405.1 | - | 3 - 4 | 395 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | mitochondrial rRNA methyltransferase 1 | MRM1 | -272.3kb | Anti-sense | Chr. 17 - 36601581 - 36634698 on NCBI Build GRCh38 | |
Downstream | LIM homeobox 1 | LHX1 |
LIM-1, LIM1 |
9.7kb | Anti-sense | Chr. 17 - 36937475 - 36944615 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
