Assay Details
| Gene Symbol: LOC105371814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135674.1 | 1 - 2 | 43 | 70 | ||
| GenBank mRNA | CX873301.1 | - | 1 - 2 | 43 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin conjugating enzyme E2 Z | UBE2Z |
HOYS7, USE1 |
Anti-sense | Chr. 17 - 48908369 - 48929060 on NCBI Build GRCh38 | |
| Downstream |
SNF8, ESCRT-II complex subunit |
SNF8 |
Dot3, EAP30, VPS22 |
21.1kb | Sense | Chr. 17 - 48930096 - 48944842 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105371814
Genomic Map