Assay Details
Gene Symbol: LOC102723727 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_135818.1 | 1 - 2 | 167 | 83 | ||
GenBank mRNA | CR740961.1 | - | 1 - 2 | 167 | 83 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC91548 | LOC91548 | -141.6kb | Sense | Chr. 1 - 211675722 - 211688044 on NCBI Build GRCh38 | |
Downstream | integrator complex subunit 7 | INTS7 |
C1orf73, INT7 |
108.9kb | Anti-sense | Chr. 1 - 211940328 - 212035660 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
