Assay Details
Gene Symbol: LOC102723727 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_135820.1 | 4 - 5 | 109 | |||
GenBank mRNA | AI218528.1 | - | 2 - 3 | 219 | 109 | |
BX110669.1 | - | 4 - 5 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000429499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000429499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000429499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000429499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000429499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000429499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000446560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000446560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000446560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000446560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000446560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000446560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000457272.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000457272.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000457272.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000457272.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000457272.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000457272.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000457272.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000457272.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000457272.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011506.hg.1 | ENST00000457272.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000429499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000429499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000429499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000429499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000429499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000429499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000430623.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000446560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000446560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000446560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000446560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000446560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000446560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000457272.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000457272.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000457272.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000457272.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000457272.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000457272.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000457272.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000457272.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000457272.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011506.hg.1 | ENST00000457272.1 | Ensembl |
