Assay Details
| Gene Symbol: LOC101929163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_136245.1 | 3 - 4 | 410 | 72 | ||
| GenBank mRNA | DB175850.1 | - | 3 - 4 | 316 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | HLA complex group 23 (non-protein coding) | HCG23 | dJ1077I5.3 | -11.8kb | Sense | Chr. 6 - 32390510 - 32393691 on NCBI Build GRCh38 |
| Downstream |
major histocompatibility complex, class II, DR alpha |
HLA-DRA |
HLA-DRA1, MLRW |
32.1kb | Sense | Chr. 6 - 32439842 - 32445046 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101929163
Genomic Map