Assay Details
| Gene Symbol: LOC101929163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_136246.1 | 1 - 2 | 247 | 122 | ||
| GenBank mRNA | BX117162.1 | - | 1 - 2 | 136 | 122 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 6 open reading frame 10 | C6orf10 | TSBP | Anti-sense | Chr. 6 - 32290523 - 32371912 on NCBI Build GRCh38 | |
| Downstream | HLA complex group 23 (non-protein coding) | HCG23 | dJ1077I5.3 | 40.5kb | Sense | Chr. 6 - 32390510 - 32393691 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101929163
Genomic Map