Assay Details
| Gene Symbol: LOC105378146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_136250.1 | 1 - 2 | 125 | 85 | ||
| GenBank mRNA | HY037713.1 | - | 1 - 2 | 89 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SPARC related modular calcium binding 2 | SMOC2 |
DTDP1, MST117, MSTP117, MSTP140, SMAP2, bA270C4A.1, bA37D8.1, dJ421D16.1 |
-289.7kb | Anti-sense | Chr. 6 - 168441151 - 168667994 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101929460 | LOC101929460 | 69.8kb | Sense | Chr. 6 - 169034171 - 169035643 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105378146
Genomic Map