Assay Details
Gene Symbol: LOC105378146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_136250.1 | 3 - 4 | 387 | 81 | ||
GenBank mRNA | HY037713.1 | - | 3 - 4 | 351 | 81 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | SPARC related modular calcium binding 2 | SMOC2 |
DTDP1, MST117, MSTP117, MSTP140, SMAP2, bA270C4A.1, bA37D8.1, dJ421D16.1 |
-259.7kb | Anti-sense | Chr. 6 - 168441151 - 168667994 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC101929460 | LOC101929460 | 80.9kb | Sense | Chr. 6 - 169034171 - 169035643 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
