Assay Details
Gene Symbol: LOC105375218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_136267.1 | 3 - 4 | 266 | 140 | ||
GenBank mRNA | KR024019.1 | - | 2 - 3 | 196 | 140 | |
KR024020.1 | - | 3 - 4 | 266 | 140 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
maturin, neural progenitor differentiation regulator homolog (Xenopus) |
MTURN |
C7orf41, Ells1 |
-84.7kb | Anti-sense | Chr. 7 - 30134936 - 30162765 on NCBI Build GRCh38 |
Downstream |
zinc and ring finger 2, E3 ubiquitin protein ligase |
ZNRF2 | RNF202 | 30.0kb | Anti-sense | Chr. 7 - 30284307 - 30367692 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
