Assay Details
Gene Symbol: LOC105379393 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_136589.1 | 1 - 2 | 233 | 62 | ||
GenBank mRNA | HY030269.1 | - | 1 - 2 | 235 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | lysine acetyltransferase 6A | KAT6A |
MOZ, MRD32, MYST-3, MYST3, RUNXBP2, ZC2HC6A, ZNF220 |
Anti-sense | Chr. 8 - 41929479 - 42052026 on NCBI Build GRCh38 | |
Downstream | adaptor related protein complex 3 mu 2 subunit | AP3M2 |
AP47B, CLA20, P47B |
48.0kb | Sense | Chr. 8 - 42152946 - 42171183 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
