Assay Details
| Gene Symbol: LINC00673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_137280.1 | 3 - 4 | 936 | 117 | ||
| NR_137281.1 | 4 - 5 | 1033 | 117 | |||
| GenBank mRNA | KR778897.1 | - | 2 - 3 | 570 | 117 | |
| KR778898.1 | - | 1 - 2 | 270 | 117 | ||
| KR778899.1 | - | 2 - 3 | 504 | 117 | ||
| KR778900.1 | - | 3 - 4 | 1036 | 117 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC146795 | LOC146795 | -48.3kb | Anti-sense | Chr. 17 - 72342811 - 72355136 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 511 | LINC00511 |
LCAL5, onco-lncRNA-12 |
177.1kb | Sense | Chr. 17 - 72598041 - 72640472 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000577773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000577773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000577773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000577828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000577828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000577828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000577828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000577828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000577828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000577828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000577828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000577828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000577828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000578206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000578206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000578206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000578206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000578206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000580175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000580175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000580175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000580175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000580948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000580948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000580948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000580948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000582712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000582712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000583460.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000583460.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000584749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000584749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000584749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | ENST00000584749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | NR_036488.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | NR_036488.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | NR_036488.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | NR_036488.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | NR_036488.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | NR_036488.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | NR_036488.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | NR_036488.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | NR_036488.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | NR_036488.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | NR_036488.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | NR_036488.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012466.hg.1 | NR_036488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000453722.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000457958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000577773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000577773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000577773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000577828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000577828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000577828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000577828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000577828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000577828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000577828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000577828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000577828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000577828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000578206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000578206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000578206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000578206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000578206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000580175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000580175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000580175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000580175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000580500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000580948.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000580948.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000580948.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000580948.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000582712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000582712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000583460.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000583460.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000584749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000584749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000584749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | ENST00000584749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | NR_036488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | NR_036488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | NR_036488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | NR_036488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | NR_036488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | NR_036488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | NR_036488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | NR_036488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | NR_036488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | NR_036488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | NR_036488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | NR_036488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012466.hg.1 | NR_036488.1 | Refseq |

LINC00673
Genomic Map