Assay Details
Gene Symbol: ZBTB49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017007835.1 | XP_016863324.1 | 3 - 4 | 1432 | 106 | |
GenBank mRNA | BC109088.1 | - | 3 - 4 | 1378 | 106 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Ly1 antibody reactive | LYAR |
ZC2HC2, ZLYAR |
-11.8kb | Anti-sense | Chr. 4 - 4267701 - 4290169 on NCBI Build GRCh38 |
Downstream | neuron specific gene family member 1 | NSG1 |
D4S234, D4S234E, NEEP21, P21 |
65.3kb | Sense | Chr. 4 - 4380988 - 4419058 on NCBI Build GRCh38 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC109088.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
cell cycle arrest
negative regulation of cell proliferation
positive regulation of transcription from RNA polymerase II promoter
transcription coactivator binding
protein binding
transcription factor binding
sequence-specific DNA binding
metal ion binding
transcription, DNA-templated
cell cycle arrest
negative regulation of cell proliferation
positive regulation of transcription from RNA polymerase II promoter
transcription coactivator binding
protein binding
transcription factor binding
sequence-specific DNA binding
metal ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000502918.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000502918.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000502918.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000504302.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000504302.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000504302.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000511458.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000511458.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000511458.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000337872.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000502918.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000502918.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000502918.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000503703.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000504302.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000504302.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000504302.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000511458.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000511458.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000511458.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | ENST00000515012.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | NM_145291.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006637.hg.1 | NM_145291.3 | Refseq |
