Assay Details
| Gene Symbol: CLLU1OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017019673.1 | XP_016875162.1 | 3 - 4 | 604 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928617 | LOC101928617 | -234.3kb | Anti-sense | Chr. 12 - 92146085 - 92185784 on NCBI Build GRCh38 | |
| Downstream | chronic lymphocytic leukemia up-regulated 1 | CLLU1 | 358.0 | Anti-sense | Chr. 12 - 92421531 - 92431002 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

CLLU1OS
Genomic Map