Assay Details
Gene Symbol: LINC00434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_047022.1 | 2 - 3 | 369 | 97 | ||
GenBank mRNA | BX103562.1 | - | 2 - 3 | 369 | 97 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | diaphanous related formin 3 | DIAPH3 |
AN, AUNA1, DIA2, DRF3, NSDAN, diap3, mDia2 |
-50.4kb | Sense | Chr. 13 - 59665583 - 60163985 on NCBI Build GRCh38 |
Downstream | tudor domain containing 3 | TDRD3 | 169.6kb | Anti-sense | Chr. 13 - 60396457 - 60573879 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
