Assay Details
Gene Symbol: MIR5571 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_049835.1 | 1 | 52 | 77 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | immunoglobulin lambda variable 4-3 | IGLV4-3 |
IGLV43, V5-1 |
-14.2kb | Sense | Chr. 22 - 22871539 - 22872035 on NCBI Build GRCh38 |
Downstream | immunoglobulin lambda like polypeptide 5 | IGLL5 |
IGL, IGLV, VL-MAR |
1.4kb | Sense | Chr. 22 - 22887780 - 22895834 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000385097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000385097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000390319.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000390319.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000390319.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000390320.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000390320.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000390321.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000390321.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000390323.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000522922.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000526893.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000526893.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000526893.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000526893.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000526893.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000526893.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000526893.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000531372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000531372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000531372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000531372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000532223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000532223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000532223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000532223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000532223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000532223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000532223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000532223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000577998.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000610541.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000610541.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000610541.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000610541.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000610541.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000610541.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000612695.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000612695.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000612695.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000612695.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000612695.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000612695.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000612695.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000613777.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000613777.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000613777.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000613777.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000613777.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000613777.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000616131.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000616131.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000616131.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000616131.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000616131.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000616131.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000617866.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000617866.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000617866.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000617866.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000617866.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000617866.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000617880.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000617880.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000617880.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000617880.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000617880.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000617880.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000620657.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000620657.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000620657.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000620657.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000620657.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000620657.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000620665.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000620665.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000620665.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000620665.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000620665.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000620665.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NM_001178126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NM_001178126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NM_001178126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NM_001178126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NM_001178126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NM_001178126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NM_001178126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NM_001256296.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NM_001256296.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NM_001256296.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NM_001256296.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NM_001256296.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NR_049835.1 | Refseq |
