Assay Details
Gene Symbol: LINC01556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_103538.1 | 1 | 426 | 68 | ||
GenBank mRNA | BX118328.1 | - | 1 | 426 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | tripartite motif containing 27 | TRIM27 |
RFP, RNF76 |
-19.8kb | Anti-sense | Chr. 6 - 28903002 - 28923991 on NCBI Build GRCh38 |
Downstream | HLA complex group 16 (non-protein coding) | HCG16 | HCG15 | 40.2kb | Sense | Chr. 6 - 28984749 - 28988536 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
