Assay Details
Gene Symbol: KRT17P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK296148.1 | - | 1 - 2 | 503 | 71 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | keratin 17 pseudogene 5 | KRT17P5 | -1.5kb | Sense | Chr. 17 - 18422181 - 18425333 on NCBI Build GRCh38 | |
Downstream | keratin 16 pseudogene 1 | KRT16P1 |
KRT14P, KRT16P |
11.4kb | Sense | Chr. 17 - 18439908 - 18442895 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700010056.hg.1 | ENST00000453795.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010056.hg.1 | ENST00000453795.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010056.hg.1 | ENST00000453795.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010056.hg.1 | ENST00000453795.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010056.hg.1 | ENST00000453795.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010056.hg.1 | ENST00000453795.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010056.hg.1 | ENST00000453795.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010056.hg.1 | ENST00000453795.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010056.hg.1 | ENST00000453795.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010056.hg.1 | ENST00000453795.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010056.hg.1 | ENST00000453795.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010056.hg.1 | ENST00000581013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010056.hg.1 | ENST00000581013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010056.hg.1 | ENST00000581013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010056.hg.1 | ENST00000581013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010056.hg.1 | ENST00000581013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010056.hg.1 | ENST00000581013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000300992.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000300992.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000300992.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000300992.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000300992.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000300992.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000300992.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000300992.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000300992.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000300992.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000326333.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000326333.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000326333.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000326333.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000326333.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000326333.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000326333.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000326333.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000326333.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000581146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000581146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000581146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000581146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000581146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000581146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000584769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000584769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000584769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000584769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000584769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000584769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012202.hg.1 | ENST00000584769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009911.hg.1 | ENST00000399211.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009911.hg.1 | ENST00000399211.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009911.hg.1 | ENST00000399211.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009911.hg.1 | ENST00000399211.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009911.hg.1 | ENST00000399211.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009911.hg.1 | ENST00000399211.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009911.hg.1 | ENST00000399211.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009911.hg.1 | ENST00000399211.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009911.hg.1 | ENST00000399211.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009911.hg.1 | ENST00000577449.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009911.hg.1 | ENST00000577449.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009911.hg.1 | ENST00000577449.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009911.hg.1 | ENST00000577449.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009911.hg.1 | ENST00000580363.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009911.hg.1 | ENST00000580363.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009911.hg.1 | ENST00000580363.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009911.hg.1 | ENST00000580363.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009911.hg.1 | ENST00000582047.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009911.hg.1 | ENST00000582047.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009911.hg.1 | ENST00000582047.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009911.hg.1 | ENST00000582047.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009911.hg.1 | ENST00000582224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009911.hg.1 | ENST00000582224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009911.hg.1 | ENST00000582224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009911.hg.1 | ENST00000582224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010056.hg.1 | ENST00000453795.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010056.hg.1 | ENST00000453795.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010056.hg.1 | ENST00000453795.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010056.hg.1 | ENST00000453795.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010056.hg.1 | ENST00000453795.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010056.hg.1 | ENST00000453795.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010056.hg.1 | ENST00000453795.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010056.hg.1 | ENST00000453795.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010056.hg.1 | ENST00000453795.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010056.hg.1 | ENST00000453795.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010056.hg.1 | ENST00000453795.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010056.hg.1 | ENST00000581013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010056.hg.1 | ENST00000581013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010056.hg.1 | ENST00000581013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010056.hg.1 | ENST00000581013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010056.hg.1 | ENST00000581013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010056.hg.1 | ENST00000581013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000300992.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000300992.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000300992.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000300992.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000300992.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000300992.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000300992.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000300992.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000300992.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000300992.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000326333.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000326333.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000326333.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000326333.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000326333.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000326333.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000326333.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000326333.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000326333.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000581146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000581146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000581146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000581146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000581146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000581146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000584769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000584769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000584769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000584769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000584769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000584769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012202.hg.1 | ENST00000584769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009911.hg.1 | ENST00000399211.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009911.hg.1 | ENST00000399211.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009911.hg.1 | ENST00000399211.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009911.hg.1 | ENST00000399211.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009911.hg.1 | ENST00000399211.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009911.hg.1 | ENST00000399211.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009911.hg.1 | ENST00000399211.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009911.hg.1 | ENST00000399211.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009911.hg.1 | ENST00000399211.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009911.hg.1 | ENST00000577449.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009911.hg.1 | ENST00000577449.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009911.hg.1 | ENST00000577449.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009911.hg.1 | ENST00000577449.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009911.hg.1 | ENST00000580363.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009911.hg.1 | ENST00000580363.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009911.hg.1 | ENST00000580363.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009911.hg.1 | ENST00000580363.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009911.hg.1 | ENST00000582047.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009911.hg.1 | ENST00000582047.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009911.hg.1 | ENST00000582047.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009911.hg.1 | ENST00000582047.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009911.hg.1 | ENST00000582224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009911.hg.1 | ENST00000582224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009911.hg.1 | ENST00000582224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009911.hg.1 | ENST00000582224.1 | Ensembl |
