Assay Details
| Gene Symbol: DDX11L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024004.1 | 1 - 2 | 121 | 99 | ||
| GenBank mRNA | AK123946.1 | - | 1 - 2 | 121 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | WAS protein family homolog 2 pseudogene | WASH2P | FAM39B | -2.2kb | Anti-sense | Chr. 2 - 113583653 - 113599037 on NCBI Build GRCh38 |
| Downstream | ribosomal protein L23a pseudogene 7 | RPL23AP7 |
RPL23AL1, RPL23A_6_267, bA395L14.9 |
7.5kb | Sense | Chr. 2 - 113611239 - 113627138 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900006432.hg.1 | ENST00000421620.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006432.hg.1 | ENST00000421620.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006432.hg.1 | ENST00000421620.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006432.hg.1 | ENST00000421620.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006432.hg.1 | ENST00000421620.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006432.hg.1 | ENST00000421620.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006432.hg.1 | ENST00000421620.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006432.hg.1 | NR_051986.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006432.hg.1 | NR_051986.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006432.hg.1 | NR_051986.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006432.hg.1 | NR_051986.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006432.hg.1 | NR_051986.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006432.hg.1 | NR_051986.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006432.hg.1 | NR_051986.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006432.hg.1 | NR_051986.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006432.hg.1 | NR_051986.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006432.hg.1 | NR_051986.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006432.hg.1 | NR_051986.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006432.hg.1 | NR_051986.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007318.hg.1 | ENSTR0000507418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007318.hg.1 | ENSTR0000507418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007318.hg.1 | ENSTR0000507418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007318.hg.1 | ENSTR0000507418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007318.hg.1 | ENSTR0000507418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011256.hg.1 | ENST00000507418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011256.hg.1 | ENST00000507418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011256.hg.1 | ENST00000507418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011256.hg.1 | ENST00000507418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011256.hg.1 | ENST00000507418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011256.hg.1 | ENST00000507418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011256.hg.1 | NR_110561.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011256.hg.1 | NR_110561.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011256.hg.1 | NR_110561.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011256.hg.1 | NR_110561.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011256.hg.1 | NR_110561.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011256.hg.1 | NR_110561.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011256.hg.1 | NR_110561.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011256.hg.1 | NR_110561.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | ENST00000450305.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | ENST00000450305.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | ENST00000450305.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | ENST00000450305.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | ENST00000450305.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | ENST00000450305.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | ENST00000456328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | ENST00000456328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | ENST00000456328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | ENST00000456328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | ENST00000456328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | ENST00000456328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | ENST00000456328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | ENST00000456328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | ENST00000456328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | ENST00000456328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | NR_046018.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | NR_046018.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | NR_046018.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | NR_046018.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | NR_046018.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | NR_046018.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | NR_046018.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | NR_046018.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | NR_046018.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006432.hg.1 | NR_046018.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011256.hg.1 | ENST00000507418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011256.hg.1 | ENST00000507418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011256.hg.1 | ENST00000507418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011256.hg.1 | ENST00000507418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011256.hg.1 | ENST00000507418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011256.hg.1 | ENST00000507418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011256.hg.1 | NR_110561.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011256.hg.1 | NR_110561.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011256.hg.1 | NR_110561.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011256.hg.1 | NR_110561.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011256.hg.1 | NR_110561.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011256.hg.1 | NR_110561.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011256.hg.1 | NR_110561.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011256.hg.1 | NR_110561.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00007318.hg.1 | ENSTR0000507418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007318.hg.1 | ENSTR0000507418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007318.hg.1 | ENSTR0000507418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007318.hg.1 | ENSTR0000507418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007318.hg.1 | ENSTR0000507418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | ENST00000450305.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | ENST00000450305.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | ENST00000450305.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | ENST00000450305.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | ENST00000450305.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | ENST00000450305.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | ENST00000456328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | ENST00000456328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | ENST00000456328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | ENST00000456328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | ENST00000456328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | ENST00000456328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | ENST00000456328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | ENST00000456328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | ENST00000456328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | ENST00000456328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | NR_046018.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | NR_046018.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | NR_046018.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | NR_046018.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | NR_046018.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | NR_046018.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | NR_046018.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | NR_046018.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | NR_046018.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006432.hg.1 | NR_046018.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006432.hg.1 | ENST00000421620.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006432.hg.1 | ENST00000421620.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006432.hg.1 | ENST00000421620.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006432.hg.1 | ENST00000421620.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006432.hg.1 | ENST00000421620.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006432.hg.1 | ENST00000421620.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006432.hg.1 | ENST00000421620.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006432.hg.1 | NR_051986.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006432.hg.1 | NR_051986.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006432.hg.1 | NR_051986.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006432.hg.1 | NR_051986.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006432.hg.1 | NR_051986.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006432.hg.1 | NR_051986.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006432.hg.1 | NR_051986.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006432.hg.1 | NR_051986.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006432.hg.1 | NR_051986.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006432.hg.1 | NR_051986.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006432.hg.1 | NR_051986.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006432.hg.1 | NR_051986.1 | Refseq |

DDX11L2
Genomic Map