Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
ENST00000554701.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
ENST00000554701.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
ENST00000554701.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
ENST00000554701.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
ENST00000554701.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
ENST00000554701.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
ENST00000554701.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
ENST00000555186.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
ENST00000555186.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
ENST00000555186.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
ENST00000555186.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
ENST00000555186.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
ENST00000555186.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
ENST00000555186.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
ENST00000566448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
ENST00000566448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
ENST00000566448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
ENST00000566448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
ENST00000566448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
ENST00000566448.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
NR_038836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
NR_038836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
NR_038836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
NR_038836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
NR_038836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
NR_038836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006447.hg.1 |
NR_038836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
ENST00000554701.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
ENST00000554701.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
ENST00000554701.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
ENST00000554701.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
ENST00000554701.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
ENST00000554701.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
ENST00000554701.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
ENST00000555186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
ENST00000555186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
ENST00000555186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
ENST00000555186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
ENST00000555186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
ENST00000555186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
ENST00000555186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
ENST00000566448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
ENST00000566448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
ENST00000566448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
ENST00000566448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
ENST00000566448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
ENST00000566448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
NR_038836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
NR_038836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
NR_038836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
NR_038836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
NR_038836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
NR_038836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006447.hg.1 |
NR_038836.1 |
Refseq |