Assay Details
Gene Symbol: ADORA2A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_028483.2 | 1 - 2 | 381 | 58 | ||
NR_028484.2 | 1 - 2 | 381 | 58 | |||
GenBank mRNA | BC045098.1 | - | 1 - 2 | 122 | 58 | |
DA917327.1 | - | 1 - 2 | 381 | 58 | ||
DB449907.2 | - | 1 - 2 | 107 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000412790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000412790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000412790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000412790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000412790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000412790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028484.3 | Refseq |
