Assay Details
| Gene Symbol: LOC100130172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037900.2 | 1 - 2 | 179 | 117 | ||
| GenBank mRNA | AK310493.1 | - | 1 - 2 | 160 | 117 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | family with sequence similarity 13 member B | FAM13B |
ARHGAP49, C5orf5, FAM13B1, KHCHP, N61 |
Anti-sense | Chr. 5 - 137937909 - 138051961 on NCBI Build GRCh38 | |
| Downstream | Wnt family member 8A | WNT8A | WNT8D | 38.4kb | Sense | Chr. 5 - 138077367 - 138092900 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC100130172
Genomic Map