Assay Details
| Gene Symbol: MIR6839 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106898.1 | 1 | 58 | 82 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100128885 | LOC100128885 | -60.5kb | Sense | Chr. 7 - 64574695 - 64618589 on NCBI Build GRCh38 | |
| Downstream | zinc finger protein 138 | ZNF138 | pHZ-32 | 115.2kb | Sense | Chr. 7 - 64794388 - 64853800 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000344930.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000344930.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000344930.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000344930.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000344930.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000344930.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000344930.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000360117.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000360117.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000360117.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000360117.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000360117.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000360117.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000360117.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000360117.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000395391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000395391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000395391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000395391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000395391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000395391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000395391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000423627.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000423627.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000423627.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000423627.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000423627.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000423627.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000423627.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000423627.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000613690.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000613690.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000613690.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000613690.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000613690.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000613690.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000613690.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000613690.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000620425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000620827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000620827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000620827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000620827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000620827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000620827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000620827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000620827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | ENST00000620827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001013746.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001013746.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001013746.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001013746.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001013746.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001013746.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001013746.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001013746.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001013746.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001013746.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001282359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001282359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001282359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001282359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001282359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001282359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001282359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001282359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001282359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001282360.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001282360.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001282360.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001282360.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001282360.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001282360.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001282360.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_001282360.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_016220.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_016220.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_016220.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_016220.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_016220.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_016220.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_016220.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_016220.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_016220.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_016220.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_016220.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NM_016220.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NR_104148.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NR_104148.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NR_104148.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NR_104148.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NR_104148.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NR_104148.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NR_104148.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NR_104149.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NR_104149.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NR_104149.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NR_104149.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NR_104149.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NR_104149.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013370.hg.1 | NR_106898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000344930.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000344930.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000344930.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000344930.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000344930.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000344930.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000344930.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000360117.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000360117.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000360117.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000360117.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000360117.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000360117.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000360117.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000360117.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000395391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000395391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000395391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000395391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000395391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000395391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000395391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000423627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000423627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000423627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000423627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000423627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000423627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000423627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000423627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000613690.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000613690.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000613690.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000613690.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000613690.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000613690.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000613690.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000613690.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000620425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000620827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000620827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000620827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000620827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000620827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000620827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000620827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000620827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | ENST00000620827.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001013746.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001013746.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001013746.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001013746.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001013746.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001013746.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001013746.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001013746.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001013746.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001013746.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001282359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001282359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001282359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001282359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001282359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001282359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001282359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001282359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001282359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001282360.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001282360.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001282360.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001282360.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001282360.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001282360.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001282360.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_001282360.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_016220.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_016220.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_016220.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_016220.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_016220.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_016220.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_016220.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_016220.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_016220.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_016220.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_016220.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NM_016220.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NR_104148.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NR_104148.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NR_104148.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NR_104148.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NR_104148.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NR_104148.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NR_104148.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NR_104149.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NR_104149.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NR_104149.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NR_104149.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NR_104149.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NR_104149.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013370.hg.1 | NR_106898.1 | Refseq |

MIR6839
Genomic Map