Assay Details
Gene Symbol: TIMM23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110767.1 | 2 - 3 | 392 | 99 | ||
GenBank mRNA | AK098044.1 | - | 2 - 3 | 377 | 99 | |
AL542628.3 | - | 2 - 3 | 379 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | poly(ADP-ribose) glycohydrolase | PARG | PARG99 | -1.2kb | Anti-sense | Chr. 10 - 49818275 - 49942027 on NCBI Build GRCh38 |
Downstream |
ArfGAP with GTPase domain, ankyrin repeat and PH domain 6 |
AGAP6 |
AGAP-6, CTGLF3, bA324H6.1 |
36.1kb | Sense | Chr. 10 - 49981196 - 50010507 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000374098.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000374098.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000374098.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000374098.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000374098.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000374098.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000374098.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000374098.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000374098.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000374098.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000374098.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000374098.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000411179.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000429104.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000429104.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000429104.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000429104.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000429104.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000429104.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000444438.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000444438.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000444438.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000444438.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000444438.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000451577.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000451577.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000451577.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000451577.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000451577.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000451577.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000451577.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000451577.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000451577.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000451577.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000478381.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000483296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000483296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000483296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000483296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000483296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000483296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000483296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000483296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000483296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | ENST00000483296.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290118.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290118.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290118.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290118.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290118.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290118.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290118.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290118.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290118.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290118.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290118.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NM_001290118.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007596.hg.1 | NR_110767.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | ENST00000580018.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | ENST00000580018.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | ENST00000580018.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | ENST00000580018.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | ENST00000580018.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | ENST00000580018.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | ENST00000580018.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | ENST00000580018.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | ENST00000580018.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | ENST00000580018.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | ENST00000580018.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NM_006327.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NM_006327.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NM_006327.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NM_006327.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NM_006327.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NM_006327.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NM_006327.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NM_006327.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NM_006327.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NM_006327.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NM_006327.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NR_073029.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NR_073029.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NR_073029.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NR_073029.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NR_073029.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NR_073029.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NR_073029.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NR_073029.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NR_073029.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NR_073029.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NR_073029.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NR_073029.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NR_073030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NR_073030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NR_073030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NR_073030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NR_073030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NR_073030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NR_073030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NR_073030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NR_073030.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007491.hg.1 | NR_073030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | ENST00000580018.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | ENST00000580018.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | ENST00000580018.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | ENST00000580018.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | ENST00000580018.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | ENST00000580018.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | ENST00000580018.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | ENST00000580018.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | ENST00000580018.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | ENST00000580018.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | ENST00000580018.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NM_006327.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NM_006327.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NM_006327.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NM_006327.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NM_006327.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NM_006327.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NM_006327.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NM_006327.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NM_006327.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NM_006327.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NM_006327.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NR_073029.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NR_073029.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NR_073029.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NR_073029.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NR_073029.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NR_073029.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NR_073029.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NR_073029.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NR_073029.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NR_073029.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NR_073029.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NR_073029.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NR_073030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NR_073030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NR_073030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NR_073030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NR_073030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NR_073030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NR_073030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NR_073030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NR_073030.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007491.hg.1 | NR_073030.1 | Refseq |
