Assay Details
Gene Symbol: TIMM23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290117.1 | NP_001277046.1 | 6 - 7 | 671 | 88 | |
NM_001290118.1 | NP_001277047.1 | 4 - 5 | 527 | 88 | ||
NR_110767.1 | 7 - 8 | 795 | 88 | |||
NR_135475.1 | 6 - 7 | 671 | 88 | |||
XM_017015460.1 | XP_016870949.1 | 6 - 7 | 623 | 88 | ||
GenBank mRNA | AK098044.1 | - | 7 - 8 | 780 | 88 | |
AK293228.1 | - | 4 - 5 | 479 | 88 | ||
AK295370.1 | - | 6 - 7 | 623 | 88 | ||
AK316490.1 | - | 6 - 7 | 623 | 88 | ||
AK316549.1 | - | 6 - 7 | 672 | 88 | ||
DA019889.1 | - | 4 - 5 | 480 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
