Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007406.hg.1 |
ENST00000580782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007406.hg.1 |
ENST00000580782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007406.hg.1 |
ENST00000580782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007406.hg.1 |
ENST00000582631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007406.hg.1 |
ENST00000582631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007406.hg.1 |
ENST00000592890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007406.hg.1 |
ENST00000592890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007406.hg.1 |
NR_110803.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007406.hg.1 |
NR_110803.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007406.hg.1 |
NR_110803.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007406.hg.1 |
ENST00000580782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007406.hg.1 |
ENST00000580782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007406.hg.1 |
ENST00000580782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007406.hg.1 |
ENST00000582631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007406.hg.1 |
ENST00000582631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007406.hg.1 |
ENST00000592890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007406.hg.1 |
ENST00000592890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007406.hg.1 |
NR_110803.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007406.hg.1 |
NR_110803.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007406.hg.1 |
NR_110803.1 |
Refseq |