Assay Details
| Gene Symbol: LOC284825 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126011.1 | 1 - 2 | 121 | 139 | ||
| GenBank mRNA | CD515480.1 | - | 1 - 2 | 121 | 139 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 314 | LINC00314 |
C21orf94, NCRNA00314 |
-67.2kb | Sense | Chr. 21 - 28013363 - 28023209 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101927973 | LOC101927973 | 14.8kb | Anti-sense | Chr. 21 - 28116094 - 28228667 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800012291.hg.1 | ENST00000517292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012291.hg.1 | ENST00000517292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000609782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000609782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000609782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000609782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000631186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000631186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000631186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000631186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012291.hg.1 | ENST00000517292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012291.hg.1 | ENST00000517292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000609782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000609782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000609782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000609782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000631186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000631186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000631186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000631186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126011.1 | Refseq |

LOC284825
Genomic Map