Assay Details
| Gene Symbol: LOC284825 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126011.1 | 2 - 3 | 171 | 106 | ||
| GenBank mRNA | CD515480.1 | - | 2 - 3 | 171 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 314 | LINC00314 |
C21orf94, NCRNA00314 |
-78.1kb | Sense | Chr. 21 - 28013363 - 28023209 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101927973 | LOC101927973 | 12.8kb | Anti-sense | Chr. 21 - 28116094 - 28228667 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC284825
Genomic Map