Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008998.hg.1 |
ENST00000580323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008998.hg.1 |
ENST00000580323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008998.hg.1 |
ENST00000580323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008998.hg.1 |
ENST00000580323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008998.hg.1 |
ENST00000583756.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008998.hg.1 |
ENST00000583756.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008998.hg.1 |
ENST00000583756.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008998.hg.1 |
ENST00000583756.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008998.hg.1 |
ENST00000583756.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008998.hg.1 |
ENST00000584810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008998.hg.1 |
ENST00000584810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008998.hg.1 |
ENST00000584810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008998.hg.1 |
NR_126324.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008998.hg.1 |
NR_126324.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008998.hg.1 |
NR_126324.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008998.hg.1 |
NR_126324.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008998.hg.1 |
NR_126324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008998.hg.1 |
ENST00000580323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008998.hg.1 |
ENST00000580323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008998.hg.1 |
ENST00000580323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008998.hg.1 |
ENST00000580323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008998.hg.1 |
ENST00000583756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008998.hg.1 |
ENST00000583756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008998.hg.1 |
ENST00000583756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008998.hg.1 |
ENST00000583756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008998.hg.1 |
ENST00000583756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008998.hg.1 |
ENST00000584810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008998.hg.1 |
ENST00000584810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008998.hg.1 |
ENST00000584810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008998.hg.1 |
NR_126324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008998.hg.1 |
NR_126324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008998.hg.1 |
NR_126324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008998.hg.1 |
NR_126324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008998.hg.1 |
NR_126324.1 |
Refseq |