Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008707.hg.1 |
ENST00000459148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013400.hg.1 |
ENST00000459201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010964.hg.1 |
ENST00000458890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008796.hg.1 |
ENST00000459407.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009981.hg.1 |
ENST00000458863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011656.hg.1 |
ENST00000521608.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011656.hg.1 |
ENST00000521608.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011656.hg.1 |
ENST00000521608.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011656.hg.1 |
ENST00000521608.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011656.hg.1 |
NR_126348.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011656.hg.1 |
NR_126348.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011656.hg.1 |
NR_126348.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011656.hg.1 |
NR_126348.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006797.hg.1 |
ENST00000458916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006797.hg.1 |
ENST00000458916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008707.hg.1 |
ENST00000459148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011656.hg.1 |
ENST00000521608.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011656.hg.1 |
ENST00000521608.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011656.hg.1 |
ENST00000521608.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011656.hg.1 |
ENST00000521608.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011656.hg.1 |
NR_126348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011656.hg.1 |
NR_126348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011656.hg.1 |
NR_126348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011656.hg.1 |
NR_126348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009981.hg.1 |
ENST00000458863.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008796.hg.1 |
ENST00000459407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013400.hg.1 |
ENST00000459201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010964.hg.1 |
ENST00000458890.1 |
Ensembl |