Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009945.hg.1 |
ENST00000446989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009945.hg.1 |
ENST00000446989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009945.hg.1 |
ENST00000446989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009945.hg.1 |
ENST00000446989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009945.hg.1 |
NR_126391.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009945.hg.1 |
NR_126391.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009945.hg.1 |
NR_126391.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009945.hg.1 |
ENST00000446989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009945.hg.1 |
ENST00000446989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009945.hg.1 |
ENST00000446989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009945.hg.1 |
ENST00000446989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009945.hg.1 |
NR_126391.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009945.hg.1 |
NR_126391.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009945.hg.1 |
NR_126391.1 |
Refseq |