| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011790.hg.1 |
ENST00000414386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011790.hg.1 |
ENST00000414386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011790.hg.1 |
ENST00000414386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011790.hg.1 |
ENST00000414386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011790.hg.1 |
ENST00000432751.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011790.hg.1 |
ENST00000432751.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011790.hg.1 |
ENST00000439386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011790.hg.1 |
ENST00000439386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011790.hg.1 |
ENST00000440194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011790.hg.1 |
ENST00000440194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011790.hg.1 |
ENST00000440194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011790.hg.1 |
ENST00000454812.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011790.hg.1 |
ENST00000454812.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011790.hg.1 |
NR_126415.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011790.hg.1 |
NR_126415.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011790.hg.1 |
NR_126416.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011790.hg.1 |
NR_126416.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011790.hg.1 |
NR_126417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011790.hg.1 |
NR_126417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011790.hg.1 |
NR_126417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011790.hg.1 |
ENST00000414386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011790.hg.1 |
ENST00000414386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011790.hg.1 |
ENST00000414386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011790.hg.1 |
ENST00000414386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011790.hg.1 |
ENST00000432751.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011790.hg.1 |
ENST00000432751.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011790.hg.1 |
ENST00000439386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011790.hg.1 |
ENST00000439386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011790.hg.1 |
ENST00000440194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011790.hg.1 |
ENST00000440194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011790.hg.1 |
ENST00000440194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011790.hg.1 |
ENST00000454812.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011790.hg.1 |
ENST00000454812.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011790.hg.1 |
NR_126415.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011790.hg.1 |
NR_126415.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011790.hg.1 |
NR_126416.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011790.hg.1 |
NR_126416.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011790.hg.1 |
NR_126417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011790.hg.1 |
NR_126417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011790.hg.1 |
NR_126417.1 |
Refseq |